Learn More
Med Vet International McKesson Sterilization Chemical Indicator Strip, 4 Inch, 73-SIS250, 2500/Case
Catalog No. 503044405
Encompass_Preferred
McKesson Sterilization Chemical Indicator Strip Steam 4 Inch For monitoring steam sterilizers operating at 250 - 285° F (121 - 140° C) The indicator strip provides distinct color change when exposed to the steam sterilization process 250 strips per box
Catalog No. 50-304-4405 Supplier Med Vet International Supplier No. 524535CS
McKesson Sterilization Chemical Indicator Strip Steam 4 Inch For monitoring steam sterilizers operating at 250 - 285° F (121 - 140° C) The indicator strip provides distinct color change when exposed to the steam sterilization process 250 strips per box