Learn More
Med Vet International 3M Attest Rapid Readout Sterilization Biological Indicator Vial, 1292, 200/Case
Catalog No. 502831955
Encompass_Preferred
Attest™ Rapid Readout Sterilization Biological Indicator Vial Steam The Attest™ steam biological indicator lets you monitor load control of sterilization and helps prevent infections The indicator features color-coding for fast referencing and ease of use The 3M™ Attest™ rapid readout biological indicator provides quick results, within 1 hour when incubated using the 3M™ Attest™ auto-reader 390 It works with 270° F/132° C gravity steam sterilizers This indicator assures sterility by challenging extremely resistant Geobacillus stearothermophilus spores Comes in a box of 50
Catalog No. 50-283-1955 Supplier Med Vet International Supplier No. 283333CS
Attest™ Rapid Readout Sterilization Biological Indicator Vial Steam The Attest™ steam biological indicator lets you monitor load control of sterilization and helps prevent infections The indicator features color-coding for fast referencing and ease of use The 3M™ Attest™ rapid readout biological indicator provides quick results, within 1 hour when incubated using the 3M™ Attest™ auto-reader 390 It works with 270° F/132° C gravity steam sterilizers This indicator assures sterility by challenging extremely resistant Geobacillus stearothermophilus spores Comes in a box of 50