Learn More
Med Vet International Attest Rapid Readout Sterilization Biological Indicator Vial Steam, 1291, 200/Case
Catalog No. 502831529
Encompass_Preferred
Attest steam biological indicator lets you monitor load control of sterilization and helps prevent infections The indicator features color-coding for fast referencing and ease of use 3M's Attest rapid readout biological indicator provides quick results, within 1 hour when incubated using 3M's Attest™ auto-reader 390 It works with 270° F gravity steam sterilizers This indicator assures sterility by challenging extremely resistant Geobacillus stearothermophilus spores
Catalog No. 50-283-1529 Supplier Med Vet International Supplier No. 196098CS
Attest steam biological indicator lets you monitor load control of sterilization and helps prevent infections The indicator features color-coding for fast referencing and ease of use 3M's Attest rapid readout biological indicator provides quick results, within 1 hour when incubated using 3M's Attest™ auto-reader 390 It works with 270° F gravity steam sterilizers This indicator assures sterility by challenging extremely resistant Geobacillus stearothermophilus spores