Learn More
Med Vet International Comply Sterilization Chemical Indicator Strip Steam 8 inch, 1250, 1920/Case
Catalog No. 502829790
Encompass_Preferred
Sterilization chemical indicator strips monitor multiple parameters Perforated in the middle of the strip to allow length to be adapted to pack size Larger portion of indicator ink for a greater area of monitoring in the pack to allow for detection of more sterilization failures
Catalog No. 50-282-9790 Supplier Med Vet International Supplier No. 181680CS
Sterilization chemical indicator strips monitor multiple parameters Perforated in the middle of the strip to allow length to be adapted to pack size Larger portion of indicator ink for a greater area of monitoring in the pack to allow for detection of more sterilization failures