Learn More
Med Vet International Comply Sterilization Chemical Indicator Strip Steam 8 inch, 1250, 240/Box
Catalog No. 502831371
Encompass_Preferred
Sterilization chemical indicator strips monitor multiple parameters Perforated in the middle of the strip to allow length to be adapted to pack size Larger portion of indicator ink for a greater area of monitoring in the pack to allow for detection of more sterilization failures
Catalog No. 50-283-1371 Supplier Med Vet International Supplier No. 181680BX
Sterilization chemical indicator strips monitor multiple parameters Perforated in the middle of the strip to allow length to be adapted to pack size Larger portion of indicator ink for a greater area of monitoring in the pack to allow for detection of more sterilization failures