Learn More
Med Vet International Attest Super Rapid Readout Sterilization Indicator Challenge Pack Steam, 1496V, 24/Case
Catalog No. 502831851
Encompass_Preferred
Challenge pack consists of multiple layers of medical index cards, some of which are die-cut to contain the biological indicator Stacked cards are wrapped with a sterilization wrap Each challenge pack has a process indicator on the pack label that changes from yellow to brown or darker when exposed to steam Meets ISO and FDA performance requirements for biological indicators Contains viable organisms that provide a direct measure of lethality
Catalog No. 50-283-1851 Supplier Med Vet International Supplier No. 852222CS
Challenge pack consists of multiple layers of medical index cards, some of which are die-cut to contain the biological indicator Stacked cards are wrapped with a sterilization wrap Each challenge pack has a process indicator on the pack label that changes from yellow to brown or darker when exposed to steam Meets ISO and FDA performance requirements for biological indicators Contains viable organisms that provide a direct measure of lethality