Learn More
Med Vet International McKesson Sterilization Chemical Indicator Strip, 8 Inch, 73-DMS250, 250/Box
Catalog No. 503043985
Encompass_Preferred
McKesson Sterilization Chemical Indicator Strip Steam / EO Gas 8 Inch Indicators change from yellow to dark brown and from yellow to orange 250 8-inch strips per box
Catalog No. 50-304-3985 Supplier Med Vet International Supplier No. 524530BX
McKesson Sterilization Chemical Indicator Strip Steam / EO Gas 8 Inch Indicators change from yellow to dark brown and from yellow to orange 250 8-inch strips per box