Learn More
Med Vet International McKesson Sterilization Chemical Indicator Strip, 8 Inch, 73-SIL250, 250/Box
Catalog No. 503044482
Encompass_Preferred
McKesson Sterilization Chemical Indicator Strip Steam 8 Inch For monitoring steam sterilizers operating at 250 - 285° F (121 - 140° C) The indicator strip provides distinct color change when exposed to the steam sterilization process 250 8-inch strips per box
Catalog No. 50-304-4482 Supplier Med Vet International Supplier No. 524534BX
 Restricted Item
Add to Cart
Add to Cart
McKesson Sterilization Chemical Indicator Strip Steam 8 Inch For monitoring steam sterilizers operating at 250 - 285° F (121 - 140° C) The indicator strip provides distinct color change when exposed to the steam sterilization process 250 8-inch strips per box