Learn More
Med Vet International Steam Indicator Strips, 4in, 250/Box
Catalog No. 502522574
Encompass_Preferred
Shows a distinctive non-reversable blue to black color change. Indicated for use in gravity and vacuum cycles. Lates-free
Catalog No. 50-252-2574 Supplier Med Vet International Supplier No. 200100
Shows a distinctive non-reversable blue to black color change. Indicated for use in gravity and vacuum cycles. Lates-free