Learn More
Sklar Instruments Indicator Tape, Sklar, 1 in. x 60 yds, Semi-Crepe Paper, Disposable, Non-Sterile. EA/1.
SDP
Catalog No. 501436929 Shop All Sklar Instruments Products
Encompass_Preferred
Indicator Tape, Sklar, 1 in. x 60 yds, Semi-Crepe Paper, Disposable, Non-Sterile. Sklar's Steam indicator tape is used for visual assurance that the package has been exposed to the sterilizing process. The color of the strip changes from baige to black during the process. This tape is 1 inch X 60 yards.
This item is not returnable. View return policy
Catalog No. 50-143-6929 Supplier Sklar Instruments Supplier No. 101132
Indicator Tape, Sklar, 1 in. x 60 yds, Semi-Crepe Paper, Disposable, Non-Sterile. Sklar's Steam indicator tape is used for visual assurance that the package has been exposed to the sterilizing process. The color of the strip changes from baige to black during the process. This tape is 1 inch X 60 yards.