Learn More
Research Products International Corp Autoclave Indicator Tape, Displays Stripes, 3/4 x 2160 Inch Roll
SDP
Catalog No. 50212741
Encompass
Autoclave indicator tape provides visual proof of sterilization.Indicator bars on the tape turn black after 15 minutes of exposure to steam heat at 120C. Tape can be marked on with pen, pencil, or marking pen after autoclaving. Adheres to paper, cloth, glass, metal and plastic. Lead free.
Catalog No. 50-212-741 Supplier Research Products International Corp Supplier No. 560150
Autoclave indicator tape provides visual proof of sterilization.Indicator bars on the tape turn black after 15 minutes of exposure to steam heat at 120C. Tape can be marked on with pen, pencil, or marking pen after autoclaving. Adheres to paper, cloth, glass, metal and plastic. Lead free.