Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indicator VB, Reagents

Catalog No. 01336180
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-180 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-180 Supplier Reagents Holdings Llc Supplier No. C84009NE100C
Add to Cart
Add to Cart

Indicator VB, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Indicator VB
CAS 64-17-5, 67-63-0, 67-63-0
Synonym Victoria Blue
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8
Color Dark Blue
Quantity 100 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.