Learn More
MSC Indicol Mini Indicol Holder, Use with Dovetail & 5/32 in Indicators
Catalog No. 5000529379
Encompass
The Indicol 400 is a Mini Indicol Holder designed for precise measurement applications. It is compatible with dovetail and 5/32 inch indicators, making it a versatile tool for various industrial and scientific settings.
  • Accessory type: Mini Indicol Holder
  • For use with dovetail and 5/32 in indicators
Catalog No. 50-005-29379 Supplier MSC Supplier No. 06451660
Add to Cart
Add to Cart
The Indicol 400 is a Mini Indicol Holder designed for precise measurement applications. It is compatible with dovetail and 5/32 inch indicators, making it a versatile tool for various industrial and scientific settings.
  • Accessory type: Mini Indicol Holder
  • For use with dovetail and 5/32 in indicators