Learn More
Cayman Chemical INDIGlo LucIfRase detect 10mL
SDP
Catalog No. 502911787
Encompass_Preferred
INDIGlo Luciferase Detection Reagent (LDR) is formulated to deliver steady, long-lived luminescence signal from mammalian reporter cells expressing firefly luciferase.
Catalog No. 50-291-1787 Supplier Cayman Chemical Supplier No. 3649510 ML
INDIGlo Luciferase Detection Reagent (LDR) is formulated to deliver steady, long-lived luminescence signal from mammalian reporter cells expressing firefly luciferase.