Learn More
Cayman Chemical INDIGlo Luciferase Detection Reagent Kit, 25 mL
SDP
Catalog No. 502911584
Encompass_Preferred
INDIGlo Luciferase Detection Reagent (LDR) is formulated to deliver steady, long-lived luminescence signal from mammalian reporter cells expressing firefly luciferase.
Catalog No. 50-291-1584 Supplier Cayman Chemical Supplier No. 3649525 ML
INDIGlo Luciferase Detection Reagent (LDR) is formulated to deliver steady, long-lived luminescence signal from mammalian reporter cells expressing firefly luciferase.