Learn More
Medchemexpress LLC HY-N0335 100mg Medchemexpress, Indigo CAS:482-89-3 Purity:>98%
SDP
Catalog No. 501933525
Encompass_Preferred
Medchemexpress, HY-N0335 100mg Indigo CAS:482-89-3 Indigo is a deep and rich color dye for indole stain, isolated from the plant Indigofera tinctoria and related species [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3525 Supplier Medchemexpress LLC Supplier No. HYN0335100MG
Medchemexpress, HY-N0335 100mg Indigo CAS:482-89-3 Indigo is a deep and rich color dye for indole stain, isolated from the plant Indigofera tinctoria and related species [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.