Learn More
Indigo Biosciences INDIGO Biosciences Assay Kit, Human FGFR4/α-Klotho, 384-well Format
Supplier: Indigo Biosciences IB43102
This assay kit is designed for the detection of human fibroblast growth factor receptor 4 / α-klotho signaling. It is provided in a 384-well format and is intended for research purposes.
- Designed for human fibroblast growth factor receptor 4 / α-klotho.
- Provided in a 384-well plate format.
- Suitable for endocrine FGF signaling studies.
- Intended for research use only, not for diagnostic or therapeutic applications.
- Mammalian reporter cells are Bio-safety Level 2.
- Stable under recommended storage conditions.