Learn More
Indigo Biosciences INDIGO Biosciences Assay Kit, Human Fibroblast Growth Factor Receptor 4, β-Klotho, 384-well Format
Supplier:  Indigo Biosciences IB43202
Encompass_Preferred
This Human Fibroblast Growth Factor Receptor 4 / β-Klotho Assay Kit is designed for research purposes, specifically for investigating endocrine FGF signaling. It utilizes mammalian reporter cells classified as Bio-safety Level 2 (BSL-2).
  • For research use only, not for diagnostic or therapeutic use in humans.
  • Utilizes mammalian reporter cells at Bio-safety Level 2.
  • Designed for endocrine FGF signaling studies.
  • Available in 384-well format assays.
Catalog No. NC3978896