Learn More
Indigo Biosciences Indigo Biosciences CHRM1 Inhibition Reporter Assay System, Human, 96-Well
Supplier:  Indigo Biosciences IB44101
Encompass_Preferred
This all-inclusive, cell-based luciferase reporter assay kit is designed for Human Cholinergic Receptor, Muscarinic 1 (CHRM1) inhibition studies. It uses proprietary mammalian cells engineered for constitutive CHRM1 expression. The kit includes CHRM1 Reporter Cells, optimized media, a reference agonist, Luciferase Detection Reagent, and a cell culture-ready 96-well assay plate, providing clear, reproducible results with exceptional cell viability post-thaw and consistent lot-to-lot performance.
  • Clear, reproducible results.
  • All-inclusive assay system.
  • Exceptional cell viability post-thaw.
  • Utilizes proprietary human cells engineered for constitutive CHRM1 expression.
  • Reporter cells prepared via CryoMite™ process for immediate dispensing.
  • Low background, high-sensitivity bio-luminescence reporter gene technology.
Catalog No. NC3978900