Learn More
Indigo Biosciences INDIGO Biosciences Human ADRB2 Reporter Assay Kit, Cell-Based, 384-well Format, All-Inclusive
Supplier:  Indigo Biosciences IB32102
Encompass_Preferred
This all-inclusive cell-based luciferase reporter assay kit is designed to target the Human Adrenoreceptor Beta 2 (ADRB2). It utilizes proprietary mammalian cells engineered for constitutive expression of ADRB2. The primary application is to screen test samples to quantify their functional activity (agonist or antagonist) against ADRB2.
  • Clear, reproducible results
  • All-inclusive assay systems
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot
Catalog No. NC3978868