Learn More
Indigo Biosciences INDIGO BIOSCIENCES Human AMY1R Reporter Assay Kit, GPCR, 96-well
Catalog No. NC3978902
Encompass_Preferred
The Human AMY1R Reporter Assay Kit is an all-inclusive cell-based luciferase reporter assay targeting the Human Amylin Type 1 Receptor (AMY1R). Its primary application is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against AMY1R.
  • Clear, reproducible results
  • All-inclusive assay systems
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot
Catalog No. NC3978902 Supplier Indigo Biosciences Supplier No. IB48101
The Human AMY1R Reporter Assay Kit is an all-inclusive cell-based luciferase reporter assay targeting the Human Amylin Type 1 Receptor (AMY1R). Its primary application is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against AMY1R.
  • Clear, reproducible results
  • All-inclusive assay systems
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot