Learn More
Indigo Biosciences INDIGO BIOSCIENCES INC HNF4α Reporter Assay System, 384-well Format
Catalog No. NC3978874
Encompass_Preferred
This nuclear receptor assay system offers a precise and sensitive method to screen compounds for inverse-agonist or agonist activities against the Human HNF4α receptor. It uses proprietary human cells in a 384-well format, leveraging bio-luminescence reporter gene technology for accurate measurements. Intended for research use only.
  • Engineered human cells express HNF4α (NR2A1) with a luciferase reporter gene.
  • Quantifies inverse-agonist/agonist activity against the Human HNF4α receptor.
  • CryoMite™ process ensures high cell viability and immediate dispensing.
  • All-inclusive kit: cryopreserved cells, media, control agonist, and detection reagents.
  • Bio-luminescence technology: low background, high sensitivity, broad dynamic range.
  • Stable light emission enables efficient batch processing.
Catalog No. NC3978874 Supplier Indigo Biosciences Supplier No. IB36002
This nuclear receptor assay system offers a precise and sensitive method to screen compounds for inverse-agonist or agonist activities against the Human HNF4α receptor. It uses proprietary human cells in a 384-well format, leveraging bio-luminescence reporter gene technology for accurate measurements. Intended for research use only.
  • Engineered human cells express HNF4α (NR2A1) with a luciferase reporter gene.
  • Quantifies inverse-agonist/agonist activity against the Human HNF4α receptor.
  • CryoMite™ process ensures high cell viability and immediate dispensing.
  • All-inclusive kit: cryopreserved cells, media, control agonist, and detection reagents.
  • Bio-luminescence technology: low background, high sensitivity, broad dynamic range.
  • Stable light emission enables efficient batch processing.