Learn More
Indigo Biosciences INDIGO BIOSCIENCES INC Human PDGFRβ Reporter Assay Kit, 1 x 96-Well Format
Catalog No. NC3978848
Encompass_Preferred
The Human PDGFRβ Reporter Assay Kit is a live cell assay that utilizes luciferase reporter technology to provide sensitive and selective quantitative measurements. It is designed for researchers studying PDGFRβ activity in a 1 x 96-well format.
  • High reproducibility and reliable quantitative measurements.
  • Results delivered within 24 hr. with a convenient “Thaw, Feed, Dose, Read” protocol.
  • Stable light emission using INDIGlo Luciferase Detection Reagent.
  • Long-term cell preservation via proprietary CryoMite™ process.
  • Reliable and accurate data from transiently transfected cells without licensing agreements.
  • Applicable in drug discovery, environmental testing, and disease state target research.
Catalog No. NC3978848 Supplier Indigo Biosciences Supplier No. IB23201
The Human PDGFRβ Reporter Assay Kit is a live cell assay that utilizes luciferase reporter technology to provide sensitive and selective quantitative measurements. It is designed for researchers studying PDGFRβ activity in a 1 x 96-well format.
  • High reproducibility and reliable quantitative measurements.
  • Results delivered within 24 hr. with a convenient “Thaw, Feed, Dose, Read” protocol.
  • Stable light emission using INDIGlo Luciferase Detection Reagent.
  • Long-term cell preservation via proprietary CryoMite™ process.
  • Reliable and accurate data from transiently transfected cells without licensing agreements.
  • Applicable in drug discovery, environmental testing, and disease state target research.