Learn More
Indigo Biosciences INDIGO BIOSCIENCES INC Human TrkA Reporter Assay Kit, 384-well Format, Human
Catalog No. NC3978849
Encompass_Preferred
This all-inclusive cell-based luciferase reporter assay kit targets Human Tropomyosin receptor kinases A (TrkA/NTRK1). It uses proprietary mammalian cells engineered for constitutive TrkA expression. The kit includes TrkA Reporter Cells, optimized media, a reference activator, Luciferase Detection Reagent, and a cell culture-ready assay plate. It quantifies functional activity (activating or inhibitory) against human TrkA or the coupled Ca+2·calcineurin / NFAT pathway. Provides clear, reproducible results, exceptional cell viability post-thaw, and consistent lot-to-lot results. Store at –80°C; do not refreeze or maintain reporter cells in extended culture.
  • Clear, reproducible results
  • All-inclusive assay systems
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot
Catalog No. NC3978849 Supplier Indigo Biosciences Supplier No. IB27012
This all-inclusive cell-based luciferase reporter assay kit targets Human Tropomyosin receptor kinases A (TrkA/NTRK1). It uses proprietary mammalian cells engineered for constitutive TrkA expression. The kit includes TrkA Reporter Cells, optimized media, a reference activator, Luciferase Detection Reagent, and a cell culture-ready assay plate. It quantifies functional activity (activating or inhibitory) against human TrkA or the coupled Ca+2·calcineurin / NFAT pathway. Provides clear, reproducible results, exceptional cell viability post-thaw, and consistent lot-to-lot results. Store at –80°C; do not refreeze or maintain reporter cells in extended culture.
  • Clear, reproducible results
  • All-inclusive assay systems
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot