Learn More
Indigo Biosciences Indigo Biosciences Reporter Assay System, Human AVPR1A, 384-well
Supplier:  Indigo Biosciences IB37002
Encompass_Preferred
This all-inclusive cell-based luciferase reporter assay kit targets the Human Arginine Vasopressin Receptor 1A (AVPR1A). It uses proprietary mammalian cells engineered for constitutive expression of AVPR1A. The kit includes reporter cells, optimized media, a reference agonist, luciferase detection reagent, and a cell culture-ready assay plate for screening test samples to quantify agonist or antagonist functional activity.
  • Clear, reproducible results
  • All-inclusive assay systems
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot
  • Utilizes proprietary human cells engineered for constitutive expression of the Human Arginine Vasopressin Receptor 1A (AVPR1A)
  • Reporter cells are cryopreserved, allowing immediate dispensing without intermediate treatment steps
Catalog No. NC3978876