Learn More
Cayman Chemical IndIgo CarmIn 25g
SDP
Catalog No. 502991083
Encompass_Preferred
An indigoid dye and a redox indicator; upon reduction, shifts from blue to yellow with absorbance maxima of 610 and 407 nm, respectively
Catalog No. 50-299-1083 Supplier Cayman Chemical Supplier No. 3152525 G
An indigoid dye and a redox indicator; upon reduction, shifts from blue to yellow with absorbance maxima of 610 and 407 nm, respectively