Learn More
Cayman Chemical IndIgo CarmIn 500g
SDP
Catalog No. 502991082
Encompass_Preferred
An indigoid dye and a redox indicator; upon reduction, shifts from blue to yellow with absorbance maxima of 610 and 407 nm, respectively
Catalog No. 50-299-1082 Supplier Cayman Chemical Supplier No. 31525500 G
An indigoid dye and a redox indicator; upon reduction, shifts from blue to yellow with absorbance maxima of 610 and 407 nm, respectively