Learn More
Cayman Chemical IndIgo CarmIn 50g
SDP
Catalog No. 502990962
Encompass_Preferred
An indigoid dye and a redox indicator; upon reduction, shifts from blue to yellow with absorbance maxima of 610 and 407 nm, respectively
Catalog No. 50-299-0962 Supplier Cayman Chemical Supplier No. 3152550 G
An indigoid dye and a redox indicator; upon reduction, shifts from blue to yellow with absorbance maxima of 610 and 407 nm, respectively