Learn More
AdipoGen Indigo Carmine
SDP
Catalog No. 502560634
Encompass_Preferred
Chemical. CAS 860-22-0. Formula C16H8N2Na2O8S2. MW 466.35. Synthetic. Indigo carmine is a blue, anionic indigoid dye consisting of an indigo dimer with sodium salts of sulfonic acids at positions 5 and 5. This compound has been used as a tissue stain, pH indicator Blue at pH11.5 to yellow at pH14.0 and tool for the determination of hypochlorite in solutions. It is also applied in detecting microorganisms, treating amyloidosis, in medical devices and drug delivery systems. It can be used as a contrast stain for plasma when red nuclear stains are used. It is widely used in food and cosmetic industry beverages, chewing gums, candies, drinks, frozen products, sweetener, tablets, sunscreen, skin and hair products and as an additive or as a dye for cationic cotton fabrics, filters, display devices, inks, toners, paints, photographic material and toys. Spectral Data Absorption lambdamax 608nm.
Catalog No. 50-256-0634 Supplier AdipoGen Supplier No. CDXI0063M100
Chemical. CAS 860-22-0. Formula C16H8N2Na2O8S2. MW 466.35. Synthetic. Indigo carmine is a blue, anionic indigoid dye consisting of an indigo dimer with sodium salts of sulfonic acids at positions 5 and 5. This compound has been used as a tissue stain, pH indicator Blue at pH11.5 to yellow at pH14.0 and tool for the determination of hypochlorite in solutions. It is also applied in detecting microorganisms, treating amyloidosis, in medical devices and drug delivery systems. It can be used as a contrast stain for plasma when red nuclear stains are used. It is widely used in food and cosmetic industry beverages, chewing gums, candies, drinks, frozen products, sweetener, tablets, sunscreen, skin and hair products and as an additive or as a dye for cationic cotton fabrics, filters, display devices, inks, toners, paints, photographic material and toys. Spectral Data Absorption lambdamax 608nm.