Learn More
Selleck Chemical LLC Indigo carmine
SDP
Catalog No. 5000003141
Encompass_Preferred
Indigo carmine (Indigotine 5 5-indigodisulfonic acid sodium salt Brilliant Indigo) is an indicator at pH 11 5-14 changing from blue to yellow It has a role as a food colouring and a histological dye
Catalog No. 50-000-03141 Supplier Selleck Chemical LLC Supplier No. S565325MG
Indigo carmine (Indigotine 5 5-indigodisulfonic acid sodium salt Brilliant Indigo) is an indicator at pH 11 5-14 changing from blue to yellow It has a role as a food colouring and a histological dye