Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indigo carmine, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015696 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5696 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5696 Supplier MilliporeSigma Supelco Supplier No. 7343625MG
Add to Cart
Add to Cart

Indigo carmine is a synthetic indigoid dye, which is generally used as a food coloring agent. It is basically used to maintain the natural color of food during processing or storage and also to improve the color, texture and appearance of food products.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 860-22-0
Physical Form Neat
Linear Formula C16H8N2 Na2O8S2
MDL Number MFCD00005723
Synonym Acid Blue 74; Indigo-5,5 ′-disulfonic acid disodium salt; Indigocarmine
Type Analytical Standard
Quantity 25 mg
Formula Weight 466.35
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.