Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Indigo Carmine, Certified, C.I. DcI-7 25 GR
SDP

Supplier:  Electron Microscopy Sciences 16910

Encompass_Preferred

(Acid Blue 74; Carmine Blue; Indigotine 1, 1A)
CAS #860-22-0 C₁₆H₈N₂O₈S₂Na₂ F.W. 466.34
Solubility: 1.3% Water; 0.0% Alcohol; 4.0% Cellosolve; 2.85% Glycol; 0.0% Xylene

Indigo Carmine is Shunway's stain for animal embryos. Used with Acid Fuchsin for staining Negri bodies. Stain Tech., 5, 34 (1928). Counterstain to Hematoxylin for staining vaginal smears. Am. J. Anat., 61, 505 (1937).

Catalog No. 50-266-54


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.