Learn More
Specifications
Specifications
| Chemical Name or Material | Indigo Carmine |
| CAS Min % | 98 |
| CAS Max % | 100 |
| Packaging | Pouch |
| Molecular Formula | C16H8N2Na2O8S2 |
| MDL Number | MFCD00005723 |
| Color | Dark Blue |
| InChI Key | KHLVKKOJDHCJMG-QDBORUFSSA-L |
| Physical Form | Powder With Coppery Luster |
| SMILES | [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
