Learn More
HIMEDIA Indigo Carmine, Hi-Cert/ACS, GRM383, Laboratory Stains and Dyes, Indicator Dye, ACS Certified, CAS 860-22-0, C.I. 73015, Redox and pH Indicator, Powder, 25 g
Catalog No. NC4013647
Encompass_Preferred
Indigo carmine, Hi-Cert/ACS is an ACS certified indicator dye (CAS 860-22-0, C.I. 73015, molecular weight 466.36) supplied as blue to dark purple crystals or powder with a copper metallic luster. A 10 ppm aqueous solution gives a clear blue solution, and the dye serves as a sensitive redox indicator, a pH indicator and a biological and analytical stain. It is also used in renal function testing and microscopy. Store below 30 C. Packaged as 25 g.
Catalog No. NC4013647 Supplier HIMEDIA Supplier No. GRM38325G
Indigo carmine, Hi-Cert/ACS is an ACS certified indicator dye (CAS 860-22-0, C.I. 73015, molecular weight 466.36) supplied as blue to dark purple crystals or powder with a copper metallic luster. A 10 ppm aqueous solution gives a clear blue solution, and the dye serves as a sensitive redox indicator, a pH indicator and a biological and analytical stain. It is also used in renal function testing and microscopy. Store below 30 C. Packaged as 25 g.