Learn More
Specifications
Specifications
| Chemical Name or Material | Indigo Stock Solution |
| Absorbance | Passes Test |
| Name Note | For Ozone |
| CAS | 68988-92-1, 7664-38-2, 67627-18-5 |
| CAS Max % | 99.82, 0.1, 0.08 |
| Identification | Passes Test |
| Packaging | Amber Poly |
| Specific Gravity | 1g/mL |
| Linear Formula | C16H7K3N2O11S3 |
| Decomposition Information | Nitrogen oxides; Carbon monoxide; Carbon dioxide; Phosphorus oxides |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.