Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indigotetrasulfonic Acid Tetrapotassium Salt Hydrate 90.0+%, TCI America™
SDP

Catalog No. I00191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I00191G Supplier TCI America Supplier No. I00191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28699-96-9
Molecular Formula C16H6K4N2O14S4
Molecular Weight (g/mol) 734.86
MDL Number MFCD00059886
InChI Key SWRJFVUVNXWCRX-UHFFFAOYSA-J
Synonym Tetrapotassium Indigotetrasulfonate
PubChem CID 16212204
IUPAC Name tetrapotassium 3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5',7,7'-tetrasulfonate
SMILES [K+].[K+].[K+].[K+].[O-]S(=O)(=O)C1=CC(=C2NC(C(=O)C2=C1)=C1NC2=C(C=C(C=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 734.86
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Indigotetrasulfonic Acid Tetrapotassium Salt Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.