Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INDICAL INC INDIMAG 2
SDP

Catalog No. NC3256778
Encompass_Preferred

  • Stand-alone benchtop instrument
  • Magnetic bead-based extraction technology
  • High-quality DNA, RNA, proteins as target analytes
  • Sample capacity of 1-48 samples per run
  • Walk-away automation for hands-free operation
  • Protocols can be pre-loaded or user-developed
  • Flexible open reagent system
  • Up to 4 washing steps supported
  • Temperature range of 4 °C to 100 °C
  • Built-in automated UV light for decontamination
  • Intuitive touchscreen user interface, no PC required
  • Dimensions of 52 cm W x 54 cm H x 66 cm D
  • Weight of 32 kg
  • Power supply of 100 – 240V/AC (+/- 10%)
  • Input frequency of 50 to 60 Hz
  • Warranty options of 1 or 2 years available
  • LED status indicator for operation status
  • USB port connectivity

Catalog No. NC3256778 Supplier INDICAL INC Supplier No. IN950048
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

  • Stand-alone benchtop instrument
  • Magnetic bead-based extraction technology
  • High-quality DNA, RNA, proteins as target analytes
  • Sample capacity of 1-48 samples per run
  • Walk-away automation for hands-free operation
  • Protocols can be pre-loaded or user-developed
  • Flexible open reagent system
  • Up to 4 washing steps supported
  • Temperature range of 4 °C to 100 °C
  • Built-in automated UV light for decontamination
  • Intuitive touchscreen user interface, no PC required
  • Dimensions of 52 cm W x 54 cm H x 66 cm D
  • Weight of 32 kg
  • Power supply of 100 – 240V/AC (+/- 10%)
  • Input frequency of 50 to 60 Hz
  • Warranty options of 1 or 2 years available
  • LED status indicator for operation status
  • USB port connectivity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.