Learn More
INDICAL INC IndiMag 2 PW Elution Set (384)
SDP
Catalog No. NC3451760
Encompass_Preferred
The IndiMag 2 PW Elution Set (384) includes 48 elution strips, racked in 4 individually vacuum-packed racks, enabling the elution of up to 384 samples. Each elution strip allows simultaneous processing of 8 samples, with volumes between 50 µl and 200 µl. Only use the elution strips provided by INDICAL, as other plasticware may not be compatible with the IndiMag 2, potentially causing operational issues. The strips are designed with ridges to ensure proper orientation, with the ridges facing down when placed in the heating block.
Catalog No. NC3451760 Supplier INDICAL INC Supplier No. PW950235
The IndiMag 2 PW Elution Set (384) includes 48 elution strips, racked in 4 individually vacuum-packed racks, enabling the elution of up to 384 samples. Each elution strip allows simultaneous processing of 8 samples, with volumes between 50 µl and 200 µl. Only use the elution strips provided by INDICAL, as other plasticware may not be compatible with the IndiMag 2, potentially causing operational issues. The strips are designed with ridges to ensure proper orientation, with the ridges facing down when placed in the heating block.