Learn More
INDICAL INC IndiMag 48 PW 8-Sample Blocks (560 reactions)
SDP
Catalog No. NC3451756
Encompass_Preferred
Plasticware (32-well blocks) for 8 x 70 samples on IndiMag 48
Catalog No. NC3451756 Supplier INDICAL INC Supplier No. PW940166
Plasticware (32-well blocks) for 8 x 70 samples on IndiMag 48