Learn More
INDICAL INC INDIMAG PATHOGEN KF96 CARTRIDGE
SDP
Catalog No. NC2038442
Encompass_Preferred
The IndiMag Pathogen KF96 Cartridge Kit is a magnetic bead-based kit designed to isolate viral nucleic acids and bacterial DNA. It simplifies high-throughput total pathogen nucleic acid extraction workflow by providing prefilled reagents cartridges with of all the necessary components (magnetic beads, washes, and elution buffer) in individual 96-well plates. The prefilled plates eliminate the need for manual reagent preparation, reduces the potential risk of user error, and minimizes the time spent on preparation. The prefilled design yields a faster and more efficient sample extraction processing on the KF96 deep-well instrument family or equivalent 96-well platforms, making it an ideal choice for laboratories looking to maximize efficiency while maintaining high-quality results.
Catalog No. NC2038442 Supplier INDICAL INC Supplier No. SP947855P196
The IndiMag Pathogen KF96 Cartridge Kit is a magnetic bead-based kit designed to isolate viral nucleic acids and bacterial DNA. It simplifies high-throughput total pathogen nucleic acid extraction workflow by providing prefilled reagents cartridges with of all the necessary components (magnetic beads, washes, and elution buffer) in individual 96-well plates. The prefilled plates eliminate the need for manual reagent preparation, reduces the potential risk of user error, and minimizes the time spent on preparation. The prefilled design yields a faster and more efficient sample extraction processing on the KF96 deep-well instrument family or equivalent 96-well platforms, making it an ideal choice for laboratories looking to maximize efficiency while maintaining high-quality results.