Learn More
INDICAL INC INDIMAG PATHOGEN KIT W/O PLAST
SDP
Catalog No. NC2058509
Encompass_Preferred
The IndiMag Pathogen Kit w/o Plastics simplifies the automated extraction of RNA and DNA from viruses, bacteria, and parasites, providing an efficient solution for total nucleic acid extraction using magnetic particle technology. Optimized for molecular pathogen detection workflows, this kit is compatible with various magnetic rod instrument systems, including the IndiMag 2, IndiMag 48(s), KingFisher Automated Purification Instruments, and other equivalent automated platforms.The kit ensures reliable, reproducible extraction of nucleic acids from a wide variety of sample types, including whole blood, serum, plasma, oral fluid, body cavity fluids (e.g., peritoneal, synovial, cerebrospinal), liquid extracts from swabs (e.g., nasal, pharyngeal, cloacal), wash fluids from bronchoalveolar lavages, urine or feces suspension, and other parasites.
Catalog No. NC2058509 Supplier INDICAL INC Supplier No. SP947257
The IndiMag Pathogen Kit w/o Plastics simplifies the automated extraction of RNA and DNA from viruses, bacteria, and parasites, providing an efficient solution for total nucleic acid extraction using magnetic particle technology. Optimized for molecular pathogen detection workflows, this kit is compatible with various magnetic rod instrument systems, including the IndiMag 2, IndiMag 48(s), KingFisher Automated Purification Instruments, and other equivalent automated platforms.The kit ensures reliable, reproducible extraction of nucleic acids from a wide variety of sample types, including whole blood, serum, plasma, oral fluid, body cavity fluids (e.g., peritoneal, synovial, cerebrospinal), liquid extracts from swabs (e.g., nasal, pharyngeal, cloacal), wash fluids from bronchoalveolar lavages, urine or feces suspension, and other parasites.