Learn More
TARGETMOL CHEMICALS INC INDIMITECAN 5MG
SDP
Catalog No. 502579642
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. IndiMitecan (LMP776) is an inhibitor of topoisomerase I (Top1) with anticancer activities [1]. purity: 98%
Catalog No. 50-257-9642 Supplier TARGETMOL CHEMICALS INC Supplier No. T602005MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. IndiMitecan (LMP776) is an inhibitor of topoisomerase I (Top1) with anticancer activities [1]. purity: 98%