Learn More
Cayman Chemical Indinavir-d6, Purity: ≥99% deuterated forms (d1-d6), CAS: 185897-02-3, 1 mg
SDP
Catalog No. 502988366
Encompass_Preferred
An internal standard for the quantification of indinavir by GC- or LC-MS
Catalog No. 50-298-8366 Supplier Cayman Chemical Supplier No. 295851 MG
An internal standard for the quantification of indinavir by GC- or LC-MS