Learn More
Medchemexpress LLC Indinavir sulfate | 157810-81-6 | 99.9% | 100 MG
SDP
Catalog No. 5000408440
Encompass_Preferred
Indinavir sulfate (MK-639) is an orally active and selective HIV-1 protease inhibitor with a Ki of 0.54 nM for PR. It demonstrates anticancer activity by inhibiting the activation of MMPs-2 hydrolysis, anti-angiogenesis, and inducing apoptosis. Indinavir sulfate is also identified as a SARS-CoV 3CLpro inhibitor.
  • Orally active and selective HIV-1 protease inhibitor
  • Inhibits activation of MMPs-2 hydrolysis
  • Exhibits anti-angiogenesis activity
  • Induces apoptosis
  • Functions as a SARS-CoV 3CLpro inhibitor
Catalog No. 50-004-08440 Supplier Medchemexpress LLC Supplier No. HYB0689A100MG
Indinavir sulfate (MK-639) is an orally active and selective HIV-1 protease inhibitor with a Ki of 0.54 nM for PR. It demonstrates anticancer activity by inhibiting the activation of MMPs-2 hydrolysis, anti-angiogenesis, and inducing apoptosis. Indinavir sulfate is also identified as a SARS-CoV 3CLpro inhibitor.
  • Orally active and selective HIV-1 protease inhibitor
  • Inhibits activation of MMPs-2 hydrolysis
  • Exhibits anti-angiogenesis activity
  • Induces apoptosis
  • Functions as a SARS-CoV 3CLpro inhibitor