Learn More
TARGETMOL CHEMICALS INC Indinavir sulfate 25MG
SDP
Catalog No. 502876066
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Indinavir sulfate (L735524 sulfate) is a HIV protease inhibitor.It inhibits the HIV-1 protease with an IC50 value of 0.41 nM and Ki value of 0.24 nM.
Catalog No. 50-287-6066 Supplier TARGETMOL CHEMICALS INC Supplier No. T586325MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Indinavir sulfate (L735524 sulfate) is a HIV protease inhibitor.It inhibits the HIV-1 protease with an IC50 value of 0.41 nM and Ki value of 0.24 nM.