Learn More
Apexbio Technology LLC Indirubin 479-41-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063086
Encompass_Preferred
Indirubin (CAS 479-41-4) is a small molecule isolated from Danggui Longhui Wan a herbal medicine formulation It exerts anti-inflammatory effects primarily through the inhibition of interferon-gamma (IFN- ) production In vitro studies demonstrated that Indirubin suppresses IFN- secretion in human bone marrow mononuclear cells (HBL-38) and murine splenocytes without affecting cell proliferation In vivo administration of Indirubin reduced ear swelling in a mouse model of 2 4 6-trinitro-1-chlorobenzene (TNCB)-induced delayed-type hypersensitivity concomitant with decreased IFN- levels in lymphocyte cultures Indirubin is under clinical investigation for conditions such as psoriasis and acute promyelocytic leukemia
Catalog No. 50-000-63086 Supplier Apexbio Technology LLC Supplier No. B36725.1
Indirubin (CAS 479-41-4) is a small molecule isolated from Danggui Longhui Wan a herbal medicine formulation It exerts anti-inflammatory effects primarily through the inhibition of interferon-gamma (IFN- ) production In vitro studies demonstrated that Indirubin suppresses IFN- secretion in human bone marrow mononuclear cells (HBL-38) and murine splenocytes without affecting cell proliferation In vivo administration of Indirubin reduced ear swelling in a mouse model of 2 4 6-trinitro-1-chlorobenzene (TNCB)-induced delayed-type hypersensitivity concomitant with decreased IFN- levels in lymphocyte cultures Indirubin is under clinical investigation for conditions such as psoriasis and acute promyelocytic leukemia