Learn More
TARGETMOL CHEMICALS INC Indirubin-3-monoxime 25MG
SDP
Catalog No. 502876735
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Indirubin-3'-monoxime (Indirubin-3'-oxime) is a potent inhibitor of GSK3beta (IC50 22 nM) and also inhibits CDKs ( (IC50s 100/180/250 nM for Cdk5/p35, Cdk1/cyclin B, Cdk2/cyclin E). Purity 99.65%
Catalog No. 50-287-6735 Supplier TARGETMOL CHEMICALS INC Supplier No. T520025MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Indirubin-3'-monoxime (Indirubin-3'-oxime) is a potent inhibitor of GSK3beta (IC50 22 nM) and also inhibits CDKs ( (IC50s 100/180/250 nM for Cdk5/p35, Cdk1/cyclin B, Cdk2/cyclin E). Purity 99.65%