Learn More
Enzo Life Sciences Indirubin-3'-monoxime (5mg). CAS: 160807-49-8
SDP
Supplier:  Enzo Life Sciences BMLCC2070005
Encompass_Preferred
A potent inhibitor of glycogen synthase kinase-3β (IC50=22 nM). Also inhibits CDK1 (p34cdc2; IC50=180 nM) and CDK5 (IC50=100 nM). It reversibly arrests asynchronous HBL-100 cells at G2. Induces apoptosis in the mammary carcinoma cell line MCF-7 (10 µM). Purity: ≥98%. Solubility: Soluble in DMSO (6mg/ml) and ethanol (4mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0407