Learn More
Selleck Chemical LLC Indirubin-3-monoxime S2009-5mg
SDP
Catalog No. 503108975
Encompass_Preferred
Indirubin-3 -monoxime (Indirubin-3 -oxime) is a selective CDK inhibitor with IC50 of 0 18 M 0 44 M 0 25 M 3 33 M 0 065 M for CDK1-cyclinB CDK2-cyclinA CDK2-cyclinE CDK4-cyclinD1 CDK5-p35 respectively Indirubin-3 -monoxime is a direct and selective 5-lipoxygenase inhibitor with IC50 of 7 8-10 M
Catalog No. 50-310-8975 Supplier Selleck Chemical LLC Supplier No. S20095MG
Indirubin-3 -monoxime (Indirubin-3 -oxime) is a selective CDK inhibitor with IC50 of 0 18 M 0 44 M 0 25 M 3 33 M 0 065 M for CDK1-cyclinB CDK2-cyclinA CDK2-cyclinE CDK4-cyclinD1 CDK5-p35 respectively Indirubin-3 -monoxime is a direct and selective 5-lipoxygenase inhibitor with IC50 of 7 8-10 M