Learn More
TARGETMOL CHEMICALS INC Indirubin-3-oxime 10MG
SDP
Catalog No. 502876443
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Indirubin-3-oxime is an effective inhibitor of cyclin-dependent protein kinases, and may play an obligate role in neuronal apoptosis in Alzheimers disease. Purity 98.26%
Catalog No. 50-287-6443 Supplier TARGETMOL CHEMICALS INC Supplier No. T913810MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Indirubin-3-oxime is an effective inhibitor of cyclin-dependent protein kinases, and may play an obligate role in neuronal apoptosis in Alzheimers disease. Purity 98.26%